Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182738.1 | NP_877402.1 | 3 - 4 | 263 | 67 | |
XM_008759745.1 | XP_008757967.1 | 4 - 5 | 441 | 67 | ||
XM_008759746.1 | XP_008757968.1 | 4 - 5 | 411 | 67 | ||
GenBank mRNA | AB086189.1 | - | 3 - 4 | 263 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |