Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107498.1 | NP_001100968.1 | 7 - 8 | 1710 | 62 | |
XM_008759143.1 | XP_008757365.1 | 8 - 9 | 1833 | 62 | ||
GenBank mRNA | BC090319.1 | - | 8 - 9 | 1648 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |