Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030987.2 | NP_112249.2 | 3 - 4 | 207 | 64 | |
XM_006239517.2 | XP_006239579.1 | 3 - 4 | 359 | 64 | ||
XM_006239518.2 | XP_006239580.1 | 3 - 4 | 359 | 64 | ||
XM_006239519.2 | XP_006239581.1 | 3 - 4 | 359 | 64 | ||
XM_006239520.2 | XP_006239582.1 | 3 - 4 | 359 | 64 | ||
XM_008764335.1 | XP_008762557.1 | 2 - 3 | 242 | 64 | ||
GenBank mRNA | AY552805.1 | - | 1 - 2 | 59 | 64 | |
BC078809.1 | - | 3 - 4 | 207 | 7123318 | 64 | |
U34958.1 | - | 1 - 2 | 59 | 64 | ||
U88324.1 | - | 1 - 2 | 96 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |