Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183328.2 | NP_899157.1 | 5 - 6 | 394 | 65 | |
XM_008762827.1 | XP_008761049.1 | 6 - 7 | 640 | 65 | ||
XM_008762828.1 | XP_008761050.1 | 6 - 7 | 515 | 65 | ||
GenBank mRNA | M94557.1 | - | 5 - 6 | 448 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |