Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035238.2 | NP_001030315.1 | 2 - 3 | 145 | 87 | |
NM_031021.3 | NP_112283.1 | 2 - 3 | 196 | 87 | ||
XM_006256081.2 | XP_006256143.1 | 2 - 3 | 232 | 87 | ||
GenBank mRNA | BC078807.1 | - | 2 - 3 | 145 | 7133064 | 87 |
CB743000.1 | - | 2 - 3 | 216 | 87 | ||
CB790784.1 | - | 2 - 3 | 242 | 87 | ||
EV778558.1 | - | 2 - 3 | 200 | 87 | ||
FQ214437.1 | - | 2 - 3 | 212 | 87 | ||
FQ224491.1 | - | 2 - 3 | 216 | 87 | ||
FQ230013.1 | - | 2 - 3 | 184 | 87 | ||
HE864425.1 | - | 1 - 2 | 80 | 87 | ||
HE864426.1 | - | 1 - 2 | 80 | 87 | ||
HE864431.1 | - | 80 | 87 | |||
HE864432.1 | - | 80 | 87 | |||
HE864433.1 | - | 80 | 87 | |||
HE864434.1 | - | 80 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |