Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130989.1 | NP_001124461.1 | 5 - 6 | 663 | 78 | |
XM_008765026.1 | XP_008763248.1 | 5 - 6 | 706 | 78 | ||
GenBank mRNA | BC166579.1 | - | 5 - 6 | 663 | 7379926 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |