Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127638.1 | NP_001121110.1 | 4 - 5 | 989 | 54 | |
XM_008769764.1 | XP_008767986.1 | 4 - 5 | 1132 | 54 | ||
GenBank mRNA | BC166443.1 | - | 4 - 5 | 989 | 8363035 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |