Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024143.2 | NP_077057.1 | 5 - 6 | 1433 | 68 | |
GenBank mRNA | AF242189.1 | - | 5 - 6 | 1434 | 68 | |
BC091147.1 | - | 5 - 6 | 1456 | 7370589 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |