Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pcdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053944.1 | NP_446396.1 | 2 - 3 | 3310 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |