Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Lca5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017484.1 | NP_001017484.1 | 2 - 3 | 105 | 71 | |
XM_006248178.2 | XP_006248240.1 | 3 - 4 | 331 | 71 | ||
XM_006248179.2 | XP_006248241.1 | 4 - 5 | 433 | 71 | ||
XM_006248180.2 | XP_006248242.1 | 3 - 4 | 380 | 71 | ||
XM_006248181.2 | XP_006248243.1 | 3 - 4 | 271 | 71 | ||
XM_006248182.2 | XP_006248244.1 | 2 - 3 | 169 | 71 | ||
GenBank mRNA | BC082082.1 | - | 2 - 3 | 105 | 7133412 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |