Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031567.1 | NP_113755.1 | 5 - 6 | 832 | 65 | |
XM_006240850.2 | XP_006240912.1 | 5 - 6 | 918 | 65 | ||
GenBank mRNA | AB012233.2 | - | 5 - 6 | 832 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |