Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109081.1 | NP_001102551.1 | 8 - 9 | 994 | 55 | |
GenBank mRNA | FQ226610.1 | - | 988 | 55 | ||
FQ227016.1 | - | 988 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |