Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasl11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002830.2 | NP_001002830.1 | 3 - 4 | 434 | 62 | |
GenBank mRNA | BC083755.1 | - | 3 - 4 | 434 | 7192690 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |