Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013874.1 | NP_001013896.1 | 2 - 3 | 222 | 75 | |
XM_006250894.2 | XP_006250956.1 | 2 - 3 | 268 | 75 | ||
GenBank mRNA | BC083718.1 | - | 2 - 3 | 222 | 7190773 | 75 |
FQ212940.1 | - | 2 - 3 | 187 | 75 | ||
FQ213030.1 | - | 2 - 3 | 231 | 75 | ||
FQ213629.1 | - | 2 - 3 | 259 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |