Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camk1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182842.1 | NP_878262.1 | 2 - 3 | 170 | 81 | |
XM_006250470.2 | XP_006250532.1 | 2 - 3 | 287 | 81 | ||
GenBank mRNA | AB101231.1 | - | 2 - 3 | 170 | 81 | |
AB101232.1 | - | 2 - 3 | 170 | 81 | ||
D86557.1 | - | 2 - 3 | 176 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Rattus norvegicus Probe spans exons |