Search Thermo Fisher Scientific
Gene Symbol: Wwc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_228858.8 | XP_228858.8 | 20 - 21 | 3517 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Probe spans exons |