Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005551.2 | NP_001005551.1 | 2 - 3 | 391 | 132 | |
XM_006256859.2 | XP_006256921.1 | 3 - 4 | 488 | 132 | ||
GenBank mRNA | BC081872.1 | - | 2 - 3 | 393 | 7106842 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |