Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tceanc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014260.2 | NP_001014282.1 | 2 - 3 | 240 | 82 | |
GenBank mRNA | BC083757.1 | - | 2 - 3 | 227 | 7192791 | 82 |
DY316983.1 | - | 2 - 3 | 240 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |