Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsnd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138979.2 | NP_620435.1 | 1 - 2 | 370 | 60 | |
GenBank mRNA | AJ421029.1 | - | 1 - 2 | 401 | 60 | |
BC081725.1 | - | 1 - 2 | 370 | 7127281 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |