Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106679.2 | NP_001100149.2 | 6 - 7 | 1309 | 86 | |
XM_006238640.2 | XP_006238702.1 | 7 - 8 | 1481 | 86 | ||
XM_006238642.2 | XP_006238704.1 | 7 - 8 | 1492 | 86 | ||
XM_006238643.2 | XP_006238705.1 | 7 - 8 | 1493 | 86 | ||
XM_006238644.2 | XP_006238706.1 | 7 - 8 | 1493 | 86 | ||
XM_006238645.2 | XP_006238707.1 | 6 - 7 | 1355 | 86 | ||
XM_008763992.1 | XP_008762214.1 | 7 - 8 | 1492 | 86 | ||
GenBank mRNA | FQ225549.1 | - | 5 - 6 | 612 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |