Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Echdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106675.1 | NP_001100145.1 | 5 - 6 | 523 | 63 | |
XM_006238507.1 | XP_006238569.1 | 3 - 4 | 454 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |