Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109317.1 | NP_001102787.1 | 1 - 2 | 459 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Recommended Assay Rattus norvegicus Probe spans exons |