Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sugp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011920.1 | NP_001011920.1 | 5 - 6 | 657 | 137 | |
GenBank mRNA | BC079341.1 | - | 5 - 6 | 657 | 7106652 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |