Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cilp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108161.1 | NP_001101631.1 | 5 - 6 | 745 | 84 | |
XM_006243259.2 | XP_006243321.1 | 6 - 7 | 900 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |