Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108600.1 | NP_001102070.1 | 7 - 8 | 1648 | 141 | |
XM_006235422.2 | XP_006235484.1 | 7 - 8 | 1698 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |