Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnttip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134400.2 | NP_599227.2 | 7 - 8 | 602 | 80 | |
XM_006235512.2 | XP_006235574.1 | 7 - 8 | 621 | 80 | ||
XM_006235513.2 | XP_006235575.1 | 7 - 8 | 570 | 80 | ||
GenBank mRNA | BC100640.1 | - | 7 - 8 | 602 | 7929876 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |