Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108267.1 | NP_001101737.1 | 6 - 7 | 1021 | 66 | |
XM_006245818.2 | XP_006245880.1 | 6 - 7 | 1777 | 66 | ||
XM_006245819.2 | XP_006245881.1 | 6 - 7 | 1476 | 66 | ||
XM_006245820.2 | XP_006245882.1 | 6 - 7 | 1913 | 66 | ||
XM_006245822.2 | XP_006245884.1 | 6 - 7 | 2603 | 66 | ||
XM_006245823.2 | XP_006245885.1 | 6 - 7 | 1437 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons |