Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Agt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134432.2 | NP_602308.1 | 2 - 3 | 1030 | 89 | |
XM_008772597.1 | XP_008770819.1 | 3 - 4 | 1230 | 89 | ||
GenBank mRNA | BC078741.1 | - | 2 - 3 | 1030 | 7128542 | 89 |
BC087679.1 | - | 1 - 2 | 866 | 7318795 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons |