Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Usp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034146.1 | NP_001029318.1 | 7 - 8 | 1522 | 67 | |
XM_008772609.1 | XP_008770831.1 | 7 - 8 | 1855 | 67 | ||
XM_008772610.1 | XP_008770832.1 | 7 - 8 | 1850 | 67 | ||
GenBank mRNA | BC105892.1 | - | 7 - 8 | 1522 | 7367339 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |