Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx3x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108246.1 | NP_001101716.1 | 2 - 3 | 184 | 115 | |
XM_006256652.2 | XP_006256714.1 | 2 - 3 | 220 | 115 | ||
GenBank mRNA | BC085914.1 | - | 2 - 3 | 189 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |