Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031769.1 | NP_113957.1 | 6 - 7 | 467 | 71 | |
GenBank mRNA | AF020346.1 | - | 6 - 7 | 467 | 71 | |
FQ213832.1 | - | 6 - 7 | 571 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |