Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Micu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199412.1 | NP_955444.1 | 9 - 10 | 1114 | 101 | |
XM_006256420.2 | XP_006256482.1 | 9 - 10 | 1229 | 101 | ||
XM_006256421.2 | XP_006256483.1 | 10 - 11 | 1222 | 101 | ||
XM_006256422.2 | XP_006256484.1 | 10 - 11 | 1216 | 101 | ||
GenBank mRNA | BC062075.1 | - | 9 - 10 | 1114 | 5599430 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |