Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007601.2 | NP_001007602.1 | 2 - 3 | 174 | 144 | |
XM_006256414.1 | XP_006256476.1 | 3 - 4 | 432 | 144 | ||
GenBank mRNA | FM060466.1 | - | 2 - 3 | 174 | 144 | |
FM067129.1 | - | 2 - 3 | 151 | 144 | ||
M83740.1 | - | 2 - 3 | 370 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |