Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008891.2 | NP_001008891.2 | 2 - 3 | 257 | 151 | |
XM_006253835.2 | XP_006253897.1 | 2 - 3 | 294 | 151 | ||
GenBank mRNA | AY321345.1 | - | 231 | 151 | ||
BC098008.1 | - | 2 - 3 | 257 | 7454159 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |