Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128183.1 | NP_001121655.1 | 2 - 3 | 247 | 100 | |
XM_006233632.2 | XP_006233694.2 | 2 - 3 | 388 | 100 | ||
GenBank mRNA | DQ537491.1 | - | 2 - 3 | 233 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |