Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749452.3 | XP_003749500.2 | 4 - 5 | 881 | 72 | |
XM_006234012.2 | XP_006234074.1 | 3 - 4 | 1043 | 72 | ||
XM_008761787.1 | XP_008760009.1 | 2 - 3 | 857 | 72 | ||
GenBank mRNA | AJ646871.1 | - | 2 - 3 | 612 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |