Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024136.1 | NP_077050.1 | 1 - 2 | 146 | 85 | |
XM_008761580.1 | XP_008759802.1 | 1 - 2 | 1211 | 85 | ||
GenBank mRNA | M12790.1 | - | 1 - 2 | 98 | 85 | |
X59832.1 | - | 1 - 2 | 138 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |