Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033236.1 | NP_150239.1 | 5 - 6 | 441 | 139 | |
GenBank mRNA | BC061542.1 | - | 5 - 6 | 483 | 6918933 | 139 |
D50694.1 | - | 5 - 6 | 441 | 139 | ||
FQ209873.1 | - | 2 - 3 | 244 | 139 | ||
FQ212821.1 | - | 5 - 6 | 471 | 139 | ||
FQ213265.1 | - | 2 - 3 | 244 | 139 | ||
FQ215767.1 | - | 2 - 3 | 244 | 139 | ||
FQ225383.1 | - | 2 - 3 | 244 | 139 | ||
FQ230973.1 | - | 5 - 6 | 500 | 139 | ||
FQ234824.1 | - | 2 - 3 | 244 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |