Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133427.3 | NP_596918.3 | 2 - 3 | 365 | 87 | |
XM_006243472.2 | XP_006243534.1 | 2 - 3 | 381 | 87 | ||
XM_008766437.1 | XP_008764659.1 | 2 - 3 | 524 | 87 | ||
GenBank mRNA | AF307840.1 | - | 1 - 2 | 124 | 87 | |
AY321370.1 | - | 1 - 2 | 124 | 87 | ||
AY321371.1 | - | 1 - 2 | 124 | 87 | ||
BC080240.1 | - | 2 - 3 | 343 | 7099950 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |