Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Traip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109004.2 | NP_001102474.1 | 12 - 13 | 1166 | 84 | |
GenBank mRNA | BC162006.1 | - | 12 - 13 | 1166 | 9089897 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |