Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012570.2 | NP_036702.1 | 3 - 4 | 741 | 107 | |
GenBank mRNA | BC081841.1 | - | 3 - 4 | 625 | 7107138 | 107 |
X14044.1 | - | 3 - 4 | 631 | 107 | ||
X14223.1 | - | 3 - 4 | 604 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |