Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191863.1 | NP_001178792.1 | 6 - 7 | 929 | 81 | |
GenBank mRNA | AF531427.1 | - | 5 - 6 | 596 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |