Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019300.1 | NP_062173.1 | 8 - 9 | 777 | 59 | |
GenBank mRNA | U67914.1 | - | 8 - 9 | 763 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Rattus norvegicus Probe spans exons |