Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: Notum | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013975.2 | NP_001013997.2 | 11 - 12 | 1288 | 59 | |
XM_006247887.2 | XP_006247949.1 | 11 - 12 | 1521 | 59 | ||
XM_006247888.2 | XP_006247950.1 | 10 - 11 | 1858 | 59 | ||
GenBank mRNA | BC090016.1 | - | 3 - 4 | 1102 | 59 | |
CO562343.1 | - | 6 - 7 | 621 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |