Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185025.1 | NP_001171954.1 | 8 - 9 | 706 | 92 | |
XM_008764655.1 | XP_008762877.1 | 8 - 9 | 661 | 92 | ||
GenBank mRNA | BF282469.1 | - | 3 - 4 | 92 | ||
FQ210674.1 | - | 8 - 9 | 724 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons |