Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105715.1 | NP_001099185.1 | 1 - 2 | 123 | 90 | |
XM_008768262.1 | XP_008766484.1 | 1 - 2 | 233 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |