Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006998.1 | NP_001006999.1 | 6 - 7 | 593 | 70 | |
XM_006230787.2 | XP_006230849.1 | 7 - 8 | 896 | 70 | ||
XM_008760132.1 | XP_008758354.1 | 6 - 7 | 1083 | 70 | ||
XM_008760133.1 | XP_008758355.1 | 6 - 7 | 1083 | 70 | ||
XM_008760134.1 | XP_008758356.1 | 6 - 7 | 577 | 70 | ||
GenBank mRNA | BC083850.1 | - | 6 - 7 | 593 | 7122451 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |