Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168633.1 | NP_001162104.1 | 1 | 2216 | 61 | |
GenBank mRNA | AF050159.1 | - | 1 | 743 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |