Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013151.2 | NP_037283.2 | 12 - 13 | 1460 | 70 | |
XM_006253347.2 | XP_006253409.1 | 12 - 13 | 1532 | 70 | ||
XM_008771348.1 | XP_008769570.1 | 13 - 14 | 1590 | 70 | ||
GenBank mRNA | BC061565.1 | - | 12 - 13 | 1460 | 6920438 | 70 |
M23697.1 | - | 11 - 12 | 1384 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |