Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012027.1 | NP_001012027.1 | 3 - 4 | 680 | 71 | |
XM_008769647.1 | XP_008767869.1 | 3 - 4 | 625 | 71 | ||
GenBank mRNA | AY321346.1 | - | 539 | 71 | ||
BC088467.1 | - | 3 - 4 | 680 | 7127992 | 71 | |
FQ209388.1 | - | 3 - 4 | 698 | 71 | ||
FQ209496.1 | - | 3 - 4 | 641 | 71 | ||
FQ209598.1 | - | 3 - 4 | 666 | 71 | ||
FQ210275.1 | - | 3 - 4 | 663 | 71 | ||
FQ210716.1 | - | 3 - 4 | 699 | 71 | ||
FQ211013.1 | - | 3 - 4 | 704 | 71 | ||
FQ211135.1 | - | 3 - 4 | 698 | 71 | ||
FQ218922.1 | - | 3 - 4 | 709 | 71 | ||
FQ218964.1 | - | 3 - 4 | 663 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |